Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuropeptide B | Npb | -2.9kb | Anti-sense | Chr. 11 - 120608477 - 120609100 on NCBI Build GRCm38 | |
| Downstream | sirtuin 7 | Sirt7 | 5.8kb | Sense | Chr. 11 - 120618372 - 120625002 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map