Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 110 | Zfp110 |
2900024E01Rik, N28112, NRIF, Nrif1 |
Sense | Chr. 7 - 12834810 - 12850584 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 31152 |
Gm31152 | 24.6kb | Anti-sense | Chr. 7 - 12864984 - 12866833 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map