Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A |
Sema6a |
9330158E07, A730020P05Rik, AI851735, Sema6A-1, Semaq, VIa, b2b997Clo |
Sense | Chr. 18 - 47245254 - 47371487 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 9130209A04 gene | 9130209A04Rik | 111.1kb | Anti-sense | Chr. 18 - 47452668 - 47470868 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map