Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | unc-5 homolog C (C. elegans) | Unc5c |
B130051O18Rik, Unc5h3, rcm |
Sense | Chr. 3 - 141465564 - 141834924 on NCBI Build GRCm38 | |
| Downstream | predicted gene 15689 | Gm15689 | OTTMUSG00000025312 | 151.7kb | Anti-sense | Chr. 3 - 141643655 - 141683463 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map