Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | U2AF homology motif (UHM) kinase 1 | Uhmk1 |
4732477C12Rik, 4930500M09Rik, AA673513, AI449218, AU021979, KIS, Kist, P-CIP2 |
Anti-sense | Chr. 1 - 170199256 - 170215898 on NCBI Build GRCm38 | |
| Downstream | SH2 domain containing 1B2 | Sh2d1b2 |
EAT-2B, Eat2b, Ert, Sh2d1c |
16.5kb | Sense | Chr. 1 - 170232871 - 170251587 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map