Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1903 | Mir1903 |
Mirn1903, mmu-mir-1903 |
-10.0kb | Sense | Chr. 8 - 128359241 - 128359320 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 32643 |
Gm32643 | 83.1kb | Anti-sense | Chr. 8 - 128453055 - 128493020 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map