Assay Details
| Gene Symbol: Gabra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK082413.1 | - | 1 | 46 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
melanoma antigen family A, 10 |
Magea10 | -99.3kb | Sense | Chr. X - 72381870 - 72386858 on NCBI Build GRCm38 | |
| Downstream | microRNA 767 | Mir767 | mmu-mir-767 | 101.0kb | Sense | Chr. X - 72589988 - 72590094 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gabra3
Genomic Map