Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | polypyrimidine tract binding protein 3 | Ptbp3 |
5830471K22Rik, AA407443, AI462022, AW107884, C86549, Rod1 |
-31.5kb | Sense | Chr. 4 - 59471868 - 59549379 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA E130308A19 gene | E130308A19Rik | 44.8kb | Anti-sense | Chr. 4 - 59626200 - 59759287 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map