Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nuclear receptor coactivator 1 | Ncoa1 |
KAT13A, SRC-1, SRC1, bHLHe74 |
Anti-sense | Chr. 12 - 4247362 - 4477210 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 31938 |
Gm31938 | 169.4kb | Sense | Chr. 12 - 4523853 - 4529122 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map