Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
glucosaminyl (N-acetyl) transferase 1, core 2 |
Gcnt1 |
5630400D21Rik, B130048E03, C2 GlcNAcT, C2GNT, IGnT |
Sense | Chr. 19 - 17326141 - 17372844 on NCBI Build GRCm38 | |
| Downstream | riboflavin kinase | Rfk |
0610038L10Rik, AF031381, KOI-4 |
37.5kb | Anti-sense | Chr. 19 - 17394043 - 17401349 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map