Assay Details
| Gene Symbol: Gm15972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090376.1 | - | 1 | 1652 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dystrobrevin alpha | Dtna |
2210407P21Rik, DTN-A, Dtn, Gm19389, a-DB-1, adbn |
-90.7kb | Anti-sense | Chr. 18 - 23310020 - 23659719 on NCBI Build GRCm38 |
| Downstream |
microtubule-associated protein, RP/EB family, member 2 |
Mapre2 |
AI314113, C820009F03Rik, D18Abb1e, EB1, EB2, RP1 |
11.0 | Anti-sense | Chr. 18 - 23752333 - 23893861 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm15972
Genomic Map