Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Janus kinase 3 | Jak3 |
fae, wil |
Sense | Chr. 8 - 71676383 - 71690577 on NCBI Build GRCm38 | |
| Downstream | UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3 | B3gnt3 | 2210008L19Rik | 1.4kb | Anti-sense | Chr. 8 - 71691373 - 71702627 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map