Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 10007 | Gm10007 | ENSMUSG00000056739 | -89.3kb | Sense | Chr. 19 - 58296204 - 58300348 on NCBI Build GRCm38 |
| Downstream | coiled-coil domain containing 172 | Ccdc172 | 1700011F14Rik | 121.6kb | Sense | Chr. 19 - 58511999 - 58553104 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map