Assay Details
| Gene Symbol: Gm31592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034176.1 | - | 5 - 6 | 685 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | thymopoietin | Tmpo |
5630400D24Rik, AI195756, AI606875, AW214352, AW547477, LAP2, TP |
-727.7kb | Anti-sense | Chr. 10 - 91147571 - 91181719 on NCBI Build GRCm38 |
| Downstream | microRNA 135a-2 | Mir135a-2 |
Mirn135a-2, mir-135a-2 |
157.4kb | Anti-sense | Chr. 10 - 92072086 - 92072185 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm31592
Genomic Map