Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cell adhesion molecule 1 | Cadm1 |
2900073G06Rik, 3100001I08Rik, AI987920, Bl2, Igsf4, Igsf4a, Necl2, RA175, RA175A, RA175B, RA175C, RA175N, ST17, SgIGSF, SynCam, Tslc1 |
Sense | Chr. 9 - 47530239 - 47853385 on NCBI Build GRCm38 | |
| Downstream | predicted gene 10677 | Gm10677 | ENSMUSG00000074363 | 233.5kb | Anti-sense | Chr. 9 - 47799160 - 47818208 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map