Assay Details
| Gene Symbol: Gm38958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131705.1 | - | 1 | 352 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 652 | Zfp652 |
9530033F24Rik, ZNF652 |
-59.2kb | Anti-sense | Chr. 11 - 95749067 - 95764715 on NCBI Build GRCm38 |
| Downstream |
phosphatase, orphan 1 |
Phospho1 |
D11Moh36, Phospo1 |
89.0 | Anti-sense | Chr. 11 - 95824500 - 95832140 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm38958
Genomic Map