Assay Details
| Gene Symbol: Gm40421 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131776.1 | - | 81 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein kinase D1 | Prkd1 |
PKD, PKD1, Pkcm, Prkcm |
-354.5kb | Sense | Chr. 12 - 50341226 - 50649248 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700008C04 gene | 1700008C04Rik | 288.0 | Anti-sense | Chr. 12 - 51005156 - 51062543 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm40421
Genomic Map