Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 6530411M01 gene | 6530411M01Rik | -386.2kb | Anti-sense | Chr. 17 - 9147719 - 9168022 on NCBI Build GRCm38 | |
| Downstream |
poly(A) binding protein, cytoplasmic 6 |
Pabpc6 |
4932702K14Rik, AI428050, Pabpc3 |
74.0kb | Anti-sense | Chr. 17 - 9666497 - 9669704 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map