Assay Details
| Gene Symbol: Gm36503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK144227.1 | - | 2 - 3 | 277 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MyoD family inhibitor domain containing | Mdfic |
Kdt1, Mdfid |
-770.1kb | Sense | Chr. 6 - 15720584 - 15802169 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 36669 |
Gm36669 | 197.7kb | Sense | Chr. 6 - 16777524 - 16788512 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm36503
Genomic Map