Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA A330069E16 gene | A330069E16Rik | -24.0kb | Anti-sense | Chr. 2 - 91237146 - 91238357 on NCBI Build GRCm38 | |
| Downstream | ADP-ribosylation factor GTPase activating protein 2 | Arfgap2 |
2310032E02Rik, Zfp289 |
2.6kb | Anti-sense | Chr. 2 - 91265115 - 91277371 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map