Assay Details
| Gene Symbol: Gpr55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131861.1 | - | 2 - 3 | 269 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4933407L21 gene | 4933407L21Rik | -41.0kb | Anti-sense | Chr. 1 - 85928483 - 85931756 on NCBI Build GRCm38 | |
| Downstream | spermatogenesis associated 3 | Spata3 |
1700011N12Rik, 1700029H01Rik, 4930424D10Rik, Mtsarg1, Tsarg1 |
42.4kb | Anti-sense | Chr. 1 - 86017840 - 86029958 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gpr55
Genomic Map