Assay Details
| Gene Symbol: Gm35835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK132154.1 | - | 4 - 5 | 799 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 125b-1 | Mir125b-1 |
Mirn125b, Mirn125b-1, mir-125b-1 |
-98.1kb | Anti-sense | Chr. 9 - 41581926 - 41582002 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 6720432D03 gene | 6720432D03Rik | 118.1kb | Sense | Chr. 9 - 41803159 - 41805683 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm35835
Genomic Map