Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA A330069E16 gene | A330069E16Rik | -15.6kb | Anti-sense | Chr. 2 - 91237146 - 91238357 on NCBI Build GRCm38 | |
| Downstream | protein kinase C and casein kinase substrate in neurons 3 | Pacsin3 |
4921507A02Rik, 6330413E15Rik, AW413130 |
99.0 | Anti-sense | Chr. 2 - 91256098 - 91264680 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map