Assay Details
| Gene Symbol: Wwp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK136811.1 | - | 1 | 3645 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | regulator of microtubule dynamics 1 | Rmdn1 |
Fam82b, RMD-1 |
-9.5kb | Sense | Chr. 4 - 19574862 - 19606932 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 7, (cationic amino acid transporter, y+ system) member 13 |
Slc7a13 |
0610009O04Rik, AGT-1, AGT1, XAT2 |
198.4kb | Sense | Chr. 4 - 19818727 - 19842213 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Wwp1
Genomic Map