Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
phosphatidylinositol-specific phospholipase C, X domain containing 3 |
Plcxd3 | B130016O10Rik | Anti-sense | Chr. 15 - 4375491 - 4575585 on NCBI Build GRCm38 | |
| Downstream | maestro heat-like repeat family member 2B | Mroh2b |
4930455B06Rik, Heatr7b2, SPIF |
63.4kb | Anti-sense | Chr. 15 - 4898234 - 4962207 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map