Assay Details
| Gene Symbol: Etv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK137368.1 | - | 1 | 5149 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
diacylglycerol kinase, beta |
Dgkb |
6430574F24, 90kda, C630029D13Rik, DAGK2, DGK, DGK-beta, mKIAA0718 |
-185.3kb | Sense | Chr. 12 - 37880168 - 38634239 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 40399 |
Gm40399 | 1.1mb | Anti-sense | Chr. 12 - 39942477 - 39950626 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Etv1
Genomic Map