Assay Details
| Gene Symbol: Gm35135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK138501.1 | - | 1 - 2 | 84 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nucleotide binding protein-like | Nubpl | 2410170E07Rik | -169.2kb | Anti-sense | Chr. 12 - 52097746 - 52310970 on NCBI Build GRCm38 |
| Downstream | Rho GTPase activating protein 5 | Arhgap5 |
p190-B, p190B |
23.2kb | Anti-sense | Chr. 12 - 52503771 - 52571978 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm35135
Genomic Map