Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | roundabout homolog 1 (Drosophila) | Robo1 |
AW494633, AW742721, DUTT1, Gm310 |
Sense | Chr. 16 - 72027553 - 73047491 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930500H12 gene | 4930500H12Rik | 254.1kb | Sense | Chr. 16 - 73072346 - 73410430 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map