Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | signal recognition particle 68 | Srp68 | 2610024I03Rik | -11.0kb | Sense | Chr. 11 - 116245166 - 116274217 on NCBI Build GRCm38 |
| Downstream | exocyst complex component 7 | Exoc7 |
Exo70, sec70 |
2.3kb | Sense | Chr. 11 - 116287998 - 116306749 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map