Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 36595 |
Gm36595 | -435.3kb | Anti-sense | Chr. 10 - 56431504 - 56475419 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930467K11 gene | 4930467K11Rik | 563.6kb | Sense | Chr. 10 - 57478383 - 57486353 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map