Assay Details
| Gene Symbol: Fabp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK163791.1 | - | 1 | 2853 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein kinase inhibitor beta, cAMP dependent, testis specific |
Pkib |
Pkibeta, Prkacn2 |
-43.8kb | Sense | Chr. 10 - 57631981 - 57741112 on NCBI Build GRCm38 |
| Downstream |
sphingomyelin phosphodiesterase, acid-like 3A |
Smpdl3a |
0610010C24Rik, AI529588, ASM3A, ASML3, ASML3A |
6.7kb | Sense | Chr. 10 - 57794544 - 57811830 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fabp7
Genomic Map