Assay Details
| Gene Symbol: Gm16998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038016.1 | 2 - 3 | 1108 | 95 | ||
| GenBank mRNA | AK138755.1 | - | 2 - 3 | 1109 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
mannosidase 1, alpha |
Man1a |
Man1a1, PCR1 |
-1.6kb | Anti-sense | Chr. 10 - 53904785 - 54076586 on NCBI Build GRCm38 |
| Downstream |
TBC1 domain family, member 32 |
Tbc1d32 |
Bromi, D630037F22Rik, b2b2284Clo |
1.9mb | Anti-sense | Chr. 10 - 56014293 - 56229456 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm16998
Genomic Map