Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | split hand/foot malformation (ectrodactyly) type 1 | Shfm1 |
DSS1, Shfdg1, Shfg |
-229.7kb | Sense | Chr. 6 - 6558275 - 6578658 on NCBI Build GRCm38 |
| Downstream |
distal-less homeobox 6, opposite strand 1 |
Dlx6os1 |
A230055N17Rik, Dlx6as1, Evf-1, Evf-2, Evf1, Evf1/2, Evf2, Shhrs, mEvf-1 |
11.5kb | Sense | Chr. 6 - 6820546 - 6869533 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map