Assay Details
| Gene Symbol: Gm30075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076338.1 | - | 3 | 1847 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 20083 |
Gm20083 | -10.3kb | Anti-sense | Chr. 7 - 73183605 - 73192432 on NCBI Build GRCm38 | |
| Downstream | uncharacterized LOC100505025 | LOC100505025 | 104.7kb | Anti-sense | Chr. 7 - 73310380 - 73315193 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm30075
Genomic Map