Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | estrogen-related receptor gamma | Esrrg |
ERR3, Errg, NR3B3, mKIAA0832 |
Sense | Chr. 1 - 187608792 - 188214884 on NCBI Build GRCm38 | |
| Downstream |
Usher syndrome 2A (autosomal recessive, mild) |
Ush2a |
A930011D15Rik, A930037M10Rik, Gm676, Gm983, Mush2a, Usherin |
229.4kb | Sense | Chr. 1 - 188262017 - 188965497 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map