Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 5086 | Gm5086 |
B230311A21, EG328314 |
-125.8kb | Anti-sense | Chr. 13 - 97560000 - 97583994 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 5330416C01 gene | 5330416C01Rik | Gm532 | 2.1kb | Sense | Chr. 13 - 97712315 - 97778952 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map