Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, H/ACA box 73a |
Snora73a |
E1, E1-7, E1b, U17A |
-153.0 | Anti-sense | Chr. 4 - 132352775 - 132352979 on NCBI Build GRCm38 |
| Downstream | phosphatase and actin regulator 4 | Phactr4 |
3110001B12Rik, AI527228, AW495572, C330013F19Rik, N28169, mKIAA4120 |
2.5kb | Anti-sense | Chr. 4 - 132355922 - 132422446 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map