Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | stem-loop binding protein | Slbp | Sense | Chr. 5 - 33640055 - 33652574 on NCBI Build GRCm38 | ||
| Downstream | transmembrane protein 129 | Tmem129 |
0610039G24Rik, 2410015J15Rik, AA409575 |
5.5kb | Sense | Chr. 5 - 33653216 - 33657911 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map