Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
TGFB-induced factor homeobox 2-like, X-linked 1 |
Tgif2lx1 |
OTTMUSG00000018465, Tex1, Tgif2lx, Tgifx1 |
-1.3mb | Sense | Chr. X - 118479708 - 118480729 on NCBI Build GRCm38 |
| Downstream |
poly(A) binding protein, cytoplasmic 5 |
Pabpc5 | C820015E17 | 177.8kb | Anti-sense | Chr. X - 119927196 - 119930165 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map