Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hemicentin 1 | Hmcn1 |
EG545370, Gm201 |
Anti-sense | Chr. 1 - 150562500 - 150993687 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA C730036E19 gene | C730036E19Rik | lncLSTR | 354.5kb | Sense | Chr. 1 - 151138034 - 151144095 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map