Assay Details
| Gene Symbol: 4933421O10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036602.1 | 2 | 2350 | 86 | ||
| GenBank mRNA | AK145080.1 | - | 2 | 2344 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Ras-related GTP binding D | Rragd |
5730543C08Rik, AI467523, C030003H22Rik, D4Ertd174e |
-6.3kb | Anti-sense | Chr. 4 - 32982998 - 33022180 on NCBI Build GRCm38 |
| Downstream | ubiquitin-conjugating enzyme E2J 1 | Ube2j1 |
0710008M05Rik, 1110030I22Rik, NCUBE-1, Ncube, Ncube1, Ubc6p |
871.0 | Anti-sense | Chr. 4 - 33031392 - 33052364 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933421O10Rik
Genomic Map