Assay Details
| Gene Symbol: Tmem29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK154711.1 | - | 2 | 3451 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 15104 | Gm15104 | OTTMUSG00000018998 | -80.2kb | Sense | Chr. X - 150311924 - 150336717 on NCBI Build GRCm38 |
| Downstream | apurinic/apyrimidinic endonuclease 2 | Apex2 |
C430040P13Rik, ape2 |
83.2kb | Sense | Chr. X - 150503629 - 150588191 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tmem29
Genomic Map