Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 137 | Mir137 |
Mirn137, mir-137, mmu-mir-137 |
-46.8kb | Sense | Chr. 3 - 118433857 - 118433929 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 26871 |
Gm26871 | 23.0kb | Sense | Chr. 3 - 118504349 - 118528369 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map