Assay Details
| Gene Symbol: Gskip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK167871.1 | - | 2 | 648 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | autophagy related 2B | Atg2b |
2410024A21Rik, AI047755, AI503411, AW558123, C030004M05Rik, C630028L02Rik, mKIAA4067 |
-4.1kb | Anti-sense | Chr. 12 - 105613536 - 105685241 on NCBI Build GRCm38 |
| Downstream | adenylate kinase 7 | Ak7 |
4930502N02Rik, AK 7 |
13.6kb | Sense | Chr. 12 - 105703859 - 105782447 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gskip
Genomic Map