Assay Details
| Gene Symbol: Gm35835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC072627.1 | - | 4 | 4349 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 125b-1 | Mir125b-1 |
Mirn125b, Mirn125b-1, mir-125b-1 |
-114.6kb | Anti-sense | Chr. 9 - 41581926 - 41582002 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 6720432D03 gene | 6720432D03Rik | 102.7kb | Sense | Chr. 9 - 41803159 - 41805683 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm35835
Genomic Map