Assay Details
| Gene Symbol: Neurl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082974.2 | NP_001076443.1 | 1 - 2 | 899 | 87 | |
| GenBank mRNA | AK004524.1 | - | 1 - 2 | 760 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Neurl2
Genomic Map