Assay Details
| Gene Symbol: Tmem29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164686.1 | NP_001158158.1 | 1 - 2 | 138 | 76 | |
| GenBank mRNA | CA526769.1 | - | 1 - 2 | 139 | 76 | |
| CK624690.1 | - | 1 - 2 | 109 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem29
Genomic Map