Assay Details
| Gene Symbol: Mrps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166031.2 | NP_001159503.2 | 2 - 3 | 141 | 94 | |
| GenBank mRNA | BB869096.1 | - | 2 - 3 | 140 | 94 | |
More Information
Set Membership: |
|

Mrps2
Genomic Map