Assay Details
| Gene Symbol: Zscan29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290819.1 | NP_001277748.1 | 2 - 3 | 77 | ||
| GenBank mRNA | BM937526.1 | - | 1 - 2 | 83 | 77 | |
More Information
Set Membership: |
|

Zscan29
Genomic Map