Assay Details
| Gene Symbol: Il1rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294171.1 | NP_001281100.1 | 1 - 2 | 58 | 70 | |
| GenBank mRNA | BY228016.1 | - | 1 - 2 | 59 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001294171.1 transcripts.
Set Membership: |
|

Il1rl1
Genomic Map